Tisk…

Ethyl-2-chloro-6-methylisonicotinate 95%

Dodavatel: Apollo Scientific

Synonyms: Ethyl 2-chloro-6-methylpyridine-4-carboxylate, Ethyl 2-chloro-6-methylisonicotinate

OR952499-1G OR952499-5G
APOSOR952499-1GEA 1820 CZK
APOSOR952499-1G APOSOR952499-5G
Ethyl-2-chloro-6-methylisonicotinate 95%
Ethyl-2-chloro-6-methylisonicotinate
Vzorec: C₉H₁₀ClNO₂
MW: 199,64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09842151
CAS číslo: 3998-88-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR