Tisk…

3-Chlorophenethyl bromide 97%

Dodavatel: Apollo Scientific

Synonyms: 3-Chlorophenethyl Bromide

OR952414-1G OR952414-25G OR952414-5G
APOSOR952414-1GEA 2860 CZK
APOSOR952414-1G APOSOR952414-25G APOSOR952414-5G
3-Chlorophenethyl bromide 97%
3-Chlorophenethyl bromide
Vzorec: C₈H₈BrCl
MW: 219.51
Storage Temperature: Ambient
MDL Number: MFCD04117465
CAS číslo: 16799-05-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR