Tisk…

(5-Methoxypyridin-3-yl)methanol 95%

Dodavatel: Apollo Scientific

OR952187-10G OR952187-1G OR952187-5G
APOSOR952187-10GEA 26450 CZK
APOSOR952187-10G APOSOR952187-1G APOSOR952187-5G
(5-Methoxypyridin-3-yl)methanol 95%
(5-Methoxypyridin-3-yl)methanol
Vzorec: C₇H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD11052844
CAS číslo: 937202-11-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR