5-Methoxy-3,4-dihydro-1,8-naphthyridin-2(1H)-one 95%
Dodavatel: Apollo Scientific
OR952165-100MG
OR952165-1G
OR952165-250MG
APOSOR952165-100MGEA
4570
CZK
APOSOR952165-100MG
APOSOR952165-1G
APOSOR952165-250MG
5-Methoxy-3,4-dihydro-1,8-naphthyridin-2(1H)-one 95%
5-Methoxy-3,4-dihydro-1,8-naphthyridin-2(1H)-one
Vzorec:
C₉H₁₀N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD11052857 CAS číslo: 1045855-18-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...