(6-Methylpyridin-3-yl)acetic acid 95%
Dodavatel: Apollo Scientific
OR952061-1G
OR952061-250MG
APOSOR952061-1GEA
8730
CZK
APOSOR952061-1G
APOSOR952061-250MG
(6-Methylpyridin-3-yl)acetic acid 95%
(6-Methylpyridin-3-yl)acetic acid
Vzorec:
C₈H₉NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00190601 CAS číslo: 19733-96-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...