Tisk…

1-Cyanocyclopentanecarboxylic acid 95%

Dodavatel: Apollo Scientific

OR952041-10G OR952041-1G OR952041-5G
APOSOR952041-10GEA 27480 CZK
APOSOR952041-10G APOSOR952041-1G APOSOR952041-5G
1-Cyanocyclopentanecarboxylic acid 95%
1-Cyanocyclopentanecarboxylic acid
Vzorec: C₇H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD07345525
CAS číslo: 540490-54-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR