5-Amino-2-ethyl-2H-indazole 96%
Dodavatel: Apollo Scientific
OR952019-1G
OR952019-5G
APOSOR952019-1GEA
1620
CZK
APOSOR952019-1G
APOSOR952019-5G
5-Amino-2-ethyl-2H-indazole 96%
5-Amino-2-ethyl-2H-indazole
Vzorec:
C₉H₁₁N₃ MW: 161,2037 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18375231 CAS číslo: 5228-52-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...