Tisk…

Isopropyl-4-hydroxypiperidine-1-carboxylate 96%

Dodavatel: Apollo Scientific

Synonyms: Isopropyl 4-hydroxypiperidine-1-carboxylate

OR952004-25G
APOSOR952004-25GEA 2820 CZK
APOSOR952004-25G
Isopropyl-4-hydroxypiperidine-1-carboxylate 96%
Isopropyl-4-hydroxypiperidine-1-carboxylate
Vzorec: C₉H₁₇NO₃
MW: 187,2400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08460168
CAS číslo: 832715-51-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR