1-(5-Chlorothien-2-yl)ethanamine 96%
Dodavatel: Apollo Scientific
OR951989-1G
OR951989-5G
APOSOR951989-1GEA
6220
CZK
APOSOR951989-1G
APOSOR951989-5G
1-(5-Chlorothien-2-yl)ethanamine 96%
1-(5-Chlorothien-2-yl)ethanamine
Vzorec:
C₆H₈ClNS MW: 161,65 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05215241 CAS číslo: 214759-44-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...