Tisk…

6-(Dimethylamino)nicotinic acid 97%

Dodavatel: Apollo Scientific

Synonyms: 2-(N,N-Dimethylamino)pyridine-5-carboxylic acid

OR951947-1G OR951947-5G
APOSOR951947-1GEA 4570 CZK
APOSOR951947-1G APOSOR951947-5G
6-(Dimethylamino)nicotinic acid 97%
6-(Dimethylamino)nicotinic acid
Vzorec: C₈H₁₀N₂O₂
MW: 166,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07357408
CAS číslo: 82846-28-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR