Tisk…

5-Ethoxypyrimidin-2-amine 95%

Dodavatel: Apollo Scientific

OR951861-1G OR951861-250MG
APOSOR951861-1GEA 6350 CZK
APOSOR951861-1G APOSOR951861-250MG
5-Ethoxypyrimidin-2-amine 95%
5-Ethoxypyrimidin-2-amine
Vzorec: C₆H₉N₃O
MW: 139.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00223638
CAS číslo: 39268-74-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR