5H-Cyclopenta[b]pyridin-7(6H)-one 98%
Dodavatel: Apollo Scientific
OR951739-1G
OR951739-250MG
APOSOR951739-1GEA
3870
CZK
APOSOR951739-1G
APOSOR951739-250MG
5H-Cyclopenta[b]pyridin-7(6H)-one 98%
5H-Cyclopenta[b]pyridin-7(6H)-one
Vzorec:
C8H7NO MW: 133,1500 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07778341 CAS číslo: 31170-78-2 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...