Tisk…

4-Chloro-1H-indole-2-carboxylic acid 95%

Dodavatel: Apollo Scientific

OR951731-1G OR951731-5G
APOSOR951731-1GEA 1330 CZK
APOSOR951731-1G APOSOR951731-5G
4-Chloro-1H-indole-2-carboxylic acid 95%
4-Chloro-1H-indole-2-carboxylic acid
Vzorec: C₉H₆ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD02664470
CAS číslo: 24621-73-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR