6-Isopropylpyridin-2-amine 98%
Dodavatel: Apollo Scientific
Synonyms:
2-Amino-6-isopropylpyridine
OR951694-1G
OR951694-5G
APOSOR951694-1GEA
2610
CZK
APOSOR951694-1G
APOSOR951694-5G
6-Isopropylpyridin-2-amine 98%
6-Isopropylpyridin-2-amine
Vzorec:
C₈H₁₂N₂ MW: 136,2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09743891 CAS číslo: 78177-12-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...