C-(1-Phenylcyclopentyl)methylamine hydrochloride 95%
Dodavatel: Apollo Scientific
Synonyms:
(1-Phenylcyclopentyl)methylamine
OR951666-10G
OR951666-1G
OR951666-5G
APOSOR951666-10GEA
21490
CZK
APOSOR951666-10G
APOSOR951666-1G
APOSOR951666-5G
C-(1-Phenylcyclopentyl)methylamine hydrochloride 95%
C-(1-Phenylcyclopentyl)methylamine hydrochloride
Vzorec:
C₁₂H₁₈ClN MW: 211,74 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01211603 CAS číslo: 17511-89-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...