Tisk…

Methyl-3-aminopicolinate 98%

Dodavatel: Apollo Scientific

Synonyms: Methyl 3-aminopyridine-2-carboxylate, Methyl 3-aminopicolinate

OR951644-25G
APOSOR951644-25GEA 10850 CZK
APOSOR951644-25G
Methyl-3-aminopicolinate 98%
Methyl-3-aminopicolinate
Vzorec: C₇H₈N₂O₂
MW: 152,1500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08695602
CAS číslo: 36052-27-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR