Tisk…
5,6,7,8-Tetrahydropyrido[4,3-d]pyrimidine hydrochloride 95%
Dodavatel: Apollo Scientific
OR951629-1G OR951629-250MG OR951629-5G
APOSOR951629-1GEA 4270 CZK
APOSOR951629-1G APOSOR951629-250MG APOSOR951629-5G
5,6,7,8-Tetrahydropyrido[4,3-d]pyrimidine hydrochloride 95%
5,6,7,8-Tetrahydropyrido[4,3-d]pyrimidine hydrochloride
Vzorec: C₇H₁₀ClN₃
Storage Temperature: Ambient
MDL Number: MFCD11043221
CAS číslo: 210538-68-4