1-Hydroxycyclohexanecarboxylic acid 95%
Dodavatel: Apollo Scientific
OR951583-5G
APOSOR951583-5GEA
3580
CZK
APOSOR951583-5G
1-Hydroxycyclohexanecarboxylic acid 95%
1-Hydroxycyclohexanecarboxylic acid
Vzorec:
C₇H₁₂O₃ MW: 144,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00045477 CAS číslo: 1123-28-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...