Pyridine-3-sulphonyl chloride 98%
Dodavatel: Apollo Scientific
OR951579-100G
OR951579-25G
APOSOR951579-100GEA
9100
CZK
APOSOR951579-100G
APOSOR951579-25G
Pyridine-3-sulphonyl chloride 98%
Pyridine-3-sulphonyl chloride
Vzorec:
C5H4ClNO2S MW: 177,6088 g/mol Hustota: 1,46 g/cm³ (25 °C) Bod vzplanutí: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03452747 CAS číslo: 16133-25-8 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...