Thieno[3,2-b]pyridin-7-ol 95%
Dodavatel: Apollo Scientific
Synonyms:
7-Hydroxythieno[3,2-b]pyridine
OR951575-5G
APOSOR951575-5GEA
1820
CZK
APOSOR951575-5G
Thieno[3,2-b]pyridin-7-ol 95%
Thieno[3,2-b]pyridin-7-ol
Vzorec:
C₇H₅NOS MW: 151,183 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01863636 CAS číslo: 107818-20-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...