5,7-Dichloro-1,6-naphthyridine 95%
Dodavatel: Apollo Scientific
OR951540-1G
OR951540-5G
APOSOR951540-1GEA
1700
CZK
APOSOR951540-1G
APOSOR951540-5G
5,7-Dichloro-1,6-naphthyridine 95%
5,7-Dichloro-1,6-naphthyridine
Vzorec:
C₈H₄Cl₂N₂ MW: 199,0368 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11518965 CAS číslo: 337958-60-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...