Tisk…

5,6,7,8-Tetrahydroisoquinoline 95%

Dodavatel: Apollo Scientific

OR951474-25G
APOSOR951474-25GEA 2980 CZK
APOSOR951474-25G
5,6,7,8-Tetrahydroisoquinoline 95%
5,6,7,8-Tetrahydroisoquinoline
Vzorec: C₉H₁₁N
MW: 133,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012168
CAS číslo: 36556-06-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR