6-Chloropyridazine-3-carbonitrile 98%
Dodavatel: Apollo Scientific
OR951394-1G
APOSOR951394-1GEA
1950
CZK
APOSOR951394-1G
6-Chloropyridazine-3-carbonitrile 98%
6-Chloropyridazine-3-carbonitrile
Vzorec:
C₅H₂ClN₃ MW: 139,5400 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09835352 CAS číslo: 35857-89-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...