Tisk…

Phenyl(pyridin-4-yl)methanamine 98%

Dodavatel: Apollo Scientific

Synonyms: C-Phenyl-c-pyridin-4-yl-methylamine

OR951385-100MG OR951385-1G OR951385-250MG
APOSOR951385-100MGEA 1370 CZK
APOSOR951385-100MG APOSOR951385-1G APOSOR951385-250MG
Phenyl(pyridin-4-yl)methanamine 98%
Phenyl(pyridin-4-yl)methanamine
Vzorec: C₁₂H₁₂N₂
MW: 184,240 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06589768
CAS číslo: 58088-57-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR