Tisk…

o,α-Dimethylbenzyl alcohol 95%

Dodavatel: Apollo Scientific

Synonyms: 2-(1-Hydroxyethyl)toluene, 2-Methylphenyl methyl carbinol, α,2-Dimethylbenzyl alcohol, 1-(2-Methylphenyl)ethanol, 1-(o-Tolyl)ethanol

OR951356-25G
APOSOR951356-25GEA 2860 CZK
APOSOR951356-25G
o,α-Dimethylbenzyl alcohol 95%
o,α-Dimethylbenzyl alcohol
Vzorec: C₉H₁₂O
MW: 136,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00046632
CAS číslo: 7287-82-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR