Tisk…

(2-Chloro-5-methylpyridin-3-yl)methanol 98%

Dodavatel: Apollo Scientific

OR951254-1G OR951254-5G
APOSOR951254-1GEA 7670 CZK
APOSOR951254-1G APOSOR951254-5G
(2-Chloro-5-methylpyridin-3-yl)methanol 98%
(2-Chloro-5-methylpyridin-3-yl)methanol
Vzorec: C₇H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD12911800
CAS číslo: 518314-64-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR