N-(2-Hydroxypyridin-3-yl)acetamide 95%
Dodavatel: Apollo Scientific
OR951209-1G
OR951209-250MG
APOSOR951209-1GEA
11580
CZK
APOSOR951209-1G
APOSOR951209-250MG
N-(2-Hydroxypyridin-3-yl)acetamide 95%
N-(2-Hydroxypyridin-3-yl)acetamide
Vzorec:
C₇H₈N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD12922754 CAS číslo: 76349-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...