Tisk…

4-Methyl-5-pyridin-4-yl-4H-[1,2,4]triazole-3-thiol

Dodavatel: Apollo Scientific
Varování

OR9512-1G OR9512-500MG OR9512-250MG
APOSOR9512-1GEA 3310 CZK
APOSOR9512-1G APOSOR9512-500MG APOSOR9512-250MG
4-Methyl-5-pyridin-4-yl-4H-[1,2,4]triazole-3-thiol
4-Methyl-5-pyridin-4-yl-4H-[1,2,4]triazole-3-thiol
Vzorec: C₈H₈N₄S
Storage Temperature: Ambient
MDL Number: MFCD00112607
CAS číslo: 3652-32-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR