Tisk…

2-(2-Isopropylphenoxy)acetic acid 95%

Dodavatel: Apollo Scientific

OR951175-1G OR951175-5G
APOSOR951175-1GEA 5530 CZK
APOSOR951175-1G APOSOR951175-5G
2-(2-Isopropylphenoxy)acetic acid 95%
2-(2-Isopropylphenoxy)acetic acid
Vzorec: C₁₁H₁₄O₃
MW: 194.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00463165
CAS číslo: 25141-58-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR