4-Methyl-5-pyridin-3-yl-4H-1,2,4-triazole-3-thiol
Dodavatel: Apollo Scientific
|
Varování
|
OR9511-1G
OR9511-500MG
APOSOR9511-1GEA
5460
CZK
APOSOR9511-1G
APOSOR9511-500MG
4-Methyl-5-pyridin-3-yl-4H-1,2,4-triazole-3-thiol
4-Methyl-5-pyridin-3-yl-4H-1,2,4-triazole-3-thiol
Vzorec:
C₈H₈N₄S MW: 192,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00798376 CAS číslo: 3652-31-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...