N-Boc-amino-(4-N-boc-piperidinyl)carboxylic acid 97%
Dodavatel: Apollo Scientific
Synonyms:
1-Boc-4-(Boc-amino)piperidine-4-carboxylic Acid
OR951024-1G
OR951024-5G
APOSOR951024-1GEA
1700
CZK
APOSOR951024-1G
APOSOR951024-5G
N-Boc-amino-(4-N-boc-piperidinyl)carboxylic acid 97%
N-Boc-amino-(4-N-boc-piperidinyl)carboxylic acid
Vzorec:
C₁₆H₂₈N₂O₆ MW: 344,408 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02683116 CAS číslo: 189321-65-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...