Tisk…

3-(4-Cyanophenoxy)propionic acid 95%

Dodavatel: Apollo Scientific

OR951012-1G OR951012-250MG
APOSOR951012-1GEA 12400 CZK
APOSOR951012-1G APOSOR951012-250MG
3-(4-Cyanophenoxy)propionic acid 95%
3-(4-Cyanophenoxy)propionic acid
Vzorec: C₁₀H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD09043034
CAS číslo: 58228-89-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR