Tisk…

4-Chloro-1,1-dimethoxybutane 97%

Dodavatel: Apollo Scientific

Synonyms: 4-Chlorobutyraldehyde dimethyl Acetal, 4-Chloro-1,1-dimethoxybutane

OR951003-25G OR951003-100G OR951003-5G
APOSOR951003-25GEA 9920 CZK
APOSOR951003-25G APOSOR951003-100G APOSOR951003-5G
4-Chloro-1,1-dimethoxybutane 97%
4-Chloro-1,1-dimethoxybutane
Vzorec: C₆H₁₃ClO₂
MW: 152,62 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09032953
CAS číslo: 29882-07-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR