2-(3-Methoxyphenoxy)propionic acid 95%
Dodavatel: Apollo Scientific
OR950987-1G
OR950987-5G
APOSOR950987-1GEA
870
CZK
APOSOR950987-1G
APOSOR950987-5G
2-(3-Methoxyphenoxy)propionic acid 95%
2-(3-Methoxyphenoxy)propionic acid
Vzorec:
C₁₀H₁₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD01931912 CAS číslo: 7309-52-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...