Tisk…

4-Chloro-2-methylpyrimidine 98%

Dodavatel: Apollo Scientific

OR950972-25G
APOSOR950972-25GEA 4400 CZK
APOSOR950972-25G
4-Chloro-2-methylpyrimidine 98%
4-Chloro-2-methylpyrimidine
Vzorec: C₅H₅ClN₂
MW: 128,5596 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00234069
CAS číslo: 4994-86-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR