Tisk…

4-Chloro-2-methylbenzaldehyde 98%

Dodavatel: Apollo Scientific

OR950969-100G OR950969-25G
APOSOR950969-100GEA 18580 CZK
APOSOR950969-100G APOSOR950969-25G
4-Chloro-2-methylbenzaldehyde 98%
4-Chloro-2-methylbenzaldehyde
Vzorec: C₈H₇ClO
Storage Temperature: Ambient
MDL Number: MFCD02261735
CAS číslo: 40137-29-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR