Tisk…

2,2'-Bipyridine-6,6'-diol 95%

Dodavatel: Apollo Scientific

Synonyms: 6,6'-Dihydroxy-2,2'-bipyridine, 6,6'-Dihydroxy-2,2'-bipyridyl

OR950887-1G
APOSOR950887-1GEA 1620 CZK
APOSOR950887-1G
2,2'-Bipyridine-6,6'-diol 95%
2,2'-Bipyridine-6,6'-diol
Vzorec: C₁₀H₈N₂O₂
MW: 188,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13194839
CAS číslo: 103505-54-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR