4-Phenylmorpholin-3-one 95%
Dodavatel: Apollo Scientific
OR950870-5G
APOSOR950870-5GEA
3440
CZK
APOSOR950870-5G
4-Phenylmorpholin-3-one 95%
4-Phenylmorpholin-3-one
Vzorec:
C₁₀H₁₁NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00624822 CAS číslo: 29518-11-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...