Tisk…

Diisopropyl fumarate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Fumaric Acid Diisopropyl Ester

OR950492-1G OR950492-5G
APOSOR950492-1GEA 622 CZK
APOSOR950492-1G APOSOR950492-5G
Diisopropyl fumarate 98%
Diisopropyl fumarate
Vzorec: C₁₀H₁₆O₄
MW: 200,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00545172
CAS číslo: 7283-70-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR