Tisk…

3-(4-Chlorophenyl)propylamine 95%

Dodavatel: Apollo Scientific

Synonyms: 3-(4-Chlorophenyl)propan-1-amine

OR950426-1G OR950426-250MG
APOSOR950426-1GEA 5590 CZK
APOSOR950426-1G APOSOR950426-250MG
3-(4-Chlorophenyl)propylamine 95%
3-(4-Chlorophenyl)propylamine
Vzorec: C₉H₁₂ClN
MW: 169,66 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06212968
CAS číslo: 18655-50-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR