Tisk…

6-Bromo-5-methyl-3-pyridineboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 6-Bromo-5-methylopyridine-3-boronic acid, 6-Bromo-5-methyl-3-pyridinylboronic acid, 2-Bromo-3-methylpyridine-5-boronic acid, (6-Bromo-5-methylpyridin-3-yl)boronic acid

OR9501-1G OR9501-5G OR9501-25G
APOSOR9501-1GEA 622 CZK
APOSOR9501-1G APOSOR9501-5G APOSOR9501-25G
6-Bromo-5-methyl-3-pyridineboronic acid
6-Bromo-5-methyl-3-pyridineboronic acid
Vzorec: C₆H₇BBrNO₂
MW: 215.84
Bod tání: 238…240 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08274468
CAS číslo: 1003043-34-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR