Tisk…

2-Chloro-3-methyl-5-pyridineboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Chloro-3-methylpyridine-5-boronic acid, 6-Chloro-5-methylpyridine-3-boronic acid, (6-Chloro-5-methylpyridin-3-yl)boronic acid

OR9500-1G OR9500-5G
APOSOR9500-1GEA 622 CZK
APOSOR9500-1G APOSOR9500-5G
2-Chloro-3-methyl-5-pyridineboronic acid
2-Chloro-3-methyl-5-pyridineboronic acid
Vzorec: C₆H₇BClNO₂
MW: 171,39 g/mol
Bod tání: 180…183 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03411564
CAS číslo: 1003043-40-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR