Tisk…

2-Ethoxy-3-pyridineboronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: (2-Ethoxypyridin-3-yl)boronic acid

OR9499-1G OR9499-25G OR9499-5G
APOSOR9499-1GEA 622 CZK
APOSOR9499-1G APOSOR9499-25G APOSOR9499-5G
2-Ethoxy-3-pyridineboronic acid
2-Ethoxy-3-pyridineboronic acid
Vzorec: C₇H₁₀BNO₃
MW: 166,97 g/mol
Bod tání: 102…104 °C
Storage Temperature: Ambient
MDL Number: MFCD06201036
CAS číslo: 854373-97-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR