Tisk…

N-Methylpiperidine-4-carboxamide 95%

Dodavatel: Apollo Scientific
Varování

OR949865-1G OR949865-250MG OR949865-5G
APOSOR949865-1GEA 2480 CZK
APOSOR949865-1G APOSOR949865-250MG APOSOR949865-5G
N-Methylpiperidine-4-carboxamide 95%
N-Methylpiperidine-4-carboxamide
Vzorec: C₇H₁₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD03411316
CAS číslo: 1903-69-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR