1,3-Dioxane-2-carboxylic acid ethyl ester 95%
Dodavatel: Apollo Scientific
OR949809-1G
OR949809-5G
APOSOR949809-1GEA
4930
CZK
APOSOR949809-1G
APOSOR949809-5G
1,3-Dioxane-2-carboxylic acid ethyl ester 95%
1,3-Dioxane-2-carboxylic acid ethyl ester
Vzorec:
C₇H₁₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD26792594 CAS číslo: 90392-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...