2-Chloro-6-isopropyl-3-pyridineboronic acid 98%
Dodavatel: Apollo Scientific
Synonyms:
2-Chloro-6-isopropylpyridine-3-boronic acid
OR9498-1G
OR9498-5G
APOSOR9498-1GEA
3150
CZK
APOSOR9498-1G
APOSOR9498-5G
2-Chloro-6-isopropyl-3-pyridineboronic acid 98%
2-Chloro-6-isopropyl-3-pyridineboronic acid
Vzorec:
C₈H₁₁BClNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD08274472 CAS číslo: 1003043-37-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...