2-Chloro-4-ethoxy-6-methylpyrimidine 95%
Dodavatel: Apollo Scientific
OR949617-1G
OR949617-250MG
APOSOR949617-1GEA
3380
CZK
APOSOR949617-1G
APOSOR949617-250MG
2-Chloro-4-ethoxy-6-methylpyrimidine 95%
2-Chloro-4-ethoxy-6-methylpyrimidine
Vzorec:
C₇H₉ClN₂O Storage Temperature: Ambient |
MDL Number:
MFCD00234338 CAS číslo: 37482-64-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...