methyl cyclobutanecarboxylate 95%
Dodavatel: Apollo Scientific
Synonyms:
Methyl cyclobutanecarboxylate
OR949536-5G
APOSOR949536-5GEA
1740
CZK
APOSOR949536-5G
methyl cyclobutanecarboxylate 95%
methyl cyclobutanecarboxylate
Vzorec:
C₆H₁₀O₂ MW: 114,144 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00095142 CAS číslo: 765-85-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...