N,N-Dimethyl-4-piperidinecarboxamide hydrochloride 97%
Dodavatel: Apollo Scientific
OR948366-1G
OR948366-500MG
OR948366-5G
APOSOR948366-1GEA
9330
CZK
APOSOR948366-1G
APOSOR948366-500MG
APOSOR948366-5G
N,N-Dimethyl-4-piperidinecarboxamide hydrochloride 97%
N,N-Dimethyl-4-piperidinecarboxamide hydrochloride
Vzorec:
C₈H₁₇ClN₂O Storage Temperature: Ambient |
MDL Number:
MFCD07781032 CAS číslo: 6270-42-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...