Tisk…

Glycidyl methyl ether 90%

Dodavatel: Apollo Scientific

Synonyms: 1,2-Epoxy-3-methoxypropane

OR947696-25G OR947696-5G OR947696-1G
APOSOR947696-25GEA 0 CZK
APOSOR947696-25G APOSOR947696-5G APOSOR947696-1G
Glycidyl methyl ether 90%
Glycidyl methyl ether
Vzorec: C₄H₈O₂
Bod vzplanutí: 25 °C
Storage Temperature: Freezer
MDL Number: MFCD00005140
CAS číslo: 930-37-0
UN: 1992
ADR: 3+6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR